Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: Molt-4.
Predicted reactivity: Mouse (86%), Rat (87%), Pig (90%), Rhesus Monkey (98%), Bovine (93%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Required for DNA recombination, repair and replication. The activity of RP-A is mediated by single-stranded DNA binding and protein interactions. Required for the efficient recruitment of the DNA double-strand break repair factor RAD51 to chromatin in response to DNA damage.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HSSB; Replication factor A protein 2; Replication protein A 32 kDa subunit; Replication protein A 32kDa subunit; Replication protein A 34 kDa subunit; Replication protein A 34kDa subunit; replication protein A2, 32kDa; replication protein A4; RF-A protein 2; RFA; RP-A p32; RP-A p34; RP4-547C9.3; RpLP1; RpP2
Gene Aliases: REPA2; RP-A p32; RP-A p34; RPA2; RPA32; RPA34; RPA4
UniProt ID: (Human) P15927
Entrez Gene ID: (Chicken) 419566, (Human) 6118
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support