Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-17166 targets RPA1 in indirect ELISA, FACS, ICC, IHC, IF and WB applications and shows reactivity with Human and Non-human primate samples.
The MA5-17166 immunogen is purified recombinant fragment of human RPA1 expressed in E. Coli.
MA5-17166 detects RPA1 which has a predicted molecular weight of approximately 68.1kDa.
This gene Plays an essential role in several cellular processes in DNA metabolism including replication, recombination and DNA repair. Binds and subsequently stabilizes single-stranded DNA intermediates and thus prevents complementary DNA from reannealing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MSTP075; Replication factor A protein 1; Replication protein A 70 kDa DNA-binding subunit; replication protein A1, 70kDa; RF-A protein 1; RP-A p70; Single-stranded DNA-binding protein
Gene Aliases: HSSB; MST075; REPA1; RF-A; RP-A; RPA1; RPA70
UniProt ID: (Human) A8K0Y9
Entrez Gene ID: (Human) 6117
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support