Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Bethyl Laboratories
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
POLR3B (RNA Polymerase III Subunit B) is a Protein Coding gene. Diseases associated with POLR3B include Leukodystrophy, Hypomyelinating, 8, With Or Without Oligodontia And/Or Hypogonadotropic Hypogonadism and Cerebellar Hypoplasia With Endosteal Sclerosis. Among its related pathways are RNA Polymerase II Transcription Initiation And Promoter Clearance and ATP/ITP metabolism. Gene Ontology (GO) annotations related to this gene include DNA-directed 5'-3' RNA polymerase activity and ribonucleoside binding. An important paralog of this gene is POLR2B.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C128; DNA-directed RNA polymerase III 127.6 kDa polypeptide; DNA-directed RNA polymerase III subunit B; DNA-directed RNA polymerase III subunit RPC2; FLJ10388; interphase nucleus and mitotic apparatus associated protein; polymerase (RNA) III (DNA directed) polypeptide B; polymerase (RNA) III subunit B; RNA polymerase III subunit C2
Gene Aliases: C128; HLD8; INMAP; POLR3B; RPC2
UniProt ID: (Human) Q9NW08
Entrez Gene ID: (Human) 55703
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support