Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: NYMDCFESNE TMGVWLHIAD KKRKKYLNNK YRTSPFNLFH HINTYEDNGF LIVDLCCWKG FEFVYNYLYL ANLRENWEEV KKNARKAPQP EVRRYVLPLN IDKADTGKNL VTLPNTTATA ILCSDETIWL EPEVLFSGPR QAFEFPQINY QKYCGKPYTY AYGLGLNHFV PDRLCKLNVK TKETWVWQEP DSYPSEPIFV SHPDALEEDD GVVLSVVVSP GAGQKPAYLL ILNAKDLSEV ARAEVEINIP VTFHGLFKKS
RPE65 is a major protein of the retinal pigment epithelium (RPE). RPE65 is essential for the regeneration of rhodopsin in the visual cycle. Mutations in RPE65 are responsible for certain forms of autosomal recessive severe retinal dystrophy, including Leber Congenital Amaurosis (LCA). Loss of function results in a block in retinoid processing required for the synthesis of 11-cis retinal and the accumulation of retinyl esters in the RPE.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: All-trans-retinyl-palmitate hydrolase; BCO family, member 3; Lutein isomerase; Meso-zeaxanthin isomerase; modifier of retinal degeneration 1; p63; RBP-binding membrane protein; retinal pigment epithelium 65-like; retinal pigment epithelium specific protein 65; retinal pigment epithelium, 65 kDa; Retinal pigment epithelium-specific 65 kDa protein; retinal pigment epithelium-specific protein 65kDa; Retinitis pigmentos; retinitis pigmentosa 20 (autosomal recessive); Retinoid isomerohydrolase; Retinol isomerase
Gene Aliases: 65kDa; A930029L06Rik; BCO3; LCA2; Mord1; mRPE65; rd12; RP20; RPE65; sRPE65
UniProt ID: (Human) Q16518, (Mouse) Q91ZQ5, (Rat) O70276
Entrez Gene ID: (Human) 6121, (Mouse) 19892, (Rat) 89826
Molecular Function:
oxygenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support