Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody can recognize all the isoforms of RPGR.
Immunogen sequence: MREPEELMP DSGAVFTFGK SKFAENNPGK FWFKNDVPVH LSCGDEHSAV VTGNNKLYMF GSNNWGQLGL GSKSAISKPT CVKALKPEKV KLAACGRNHT LVSTEGGNVY ATGGNNEGQL GLGDTEERNT FHVISFFTSE HKIKQLSAGS NTSAALTEDG RLFMWGDNSE GQIGLKNVSN VCVPQQVTIG KPVSWISCGY YHSAFVTTDG ELYVFGEPEN GKLGLPNQLL GNHRTPQLVS EIPEKVIQVA CGGEHTVVLT ENAVYTFGLG QFGQLGLGTF LFETSEPKVI ENIRDQTISY ISCGENHTAL ITDIGLMYTF GDGRHGKLGL GLENFTNHFI PTLCSNFLRF IVKLVACGGC (1-359 aa encoded by BC031624)
This gene encodes a protein with a series of six RCC1-like domains (RLDs), characteristic of the highly conserved guanine nucleotide exchange factors. The encoded protein is found in the Golgi body and interacts with RPGRIP1. This protein localizes to the outer segment of rod photoreceptors and is essential for their viability. Mutations in this gene have been associated with X-linked retinitis pigmentosa (XLRP). Multiple alternatively spliced transcript variants that encode different isoforms of this gene have been reported, but the full-length natures of only some have been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mRpgr; retinitis pigmentosa 15; retinitis pigmentosa 3 GTPase regulator; X-linked retinitis pigmentosa GTPase regulator
Gene Aliases: COD1; CORDX1; CRD; orf15; PCDX; RP15; RP3; Rp3h; RPGR; XLRP3
UniProt ID: (Human) Q92834, (Mouse) Q9R0X5
Entrez Gene ID: (Human) 6103, (Mouse) 19893
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support