Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
The protein encoded by this gene can localize to the basal body-centrosome complex or to primary cilia and centrosomes in ciliated cells. The encoded protein has been found to interact with nephrocystin-4. Defects in this gene are a cause of Joubert syndrome type 7 and Meckel syndrome type 5. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686C0668; fantom; fantom homolog; Nephrocystin-8; Protein fantom; protein phosphatase 1, regulatory subunit 134; RPGR-interacting protein 1-like protein; RPGRIP1-like; RPGRIP1-like protein
Gene Aliases: 1700047E16Rik; 4931437C01; CORS3; FTM; JBTS7; KIAA1005; MKS5; NPHP8; PPP1R134; RGD1311099; RPGRIP1L
UniProt ID: (Human) Q68CZ1, (Mouse) Q8CG73
Entrez Gene ID: (Human) 23322, (Mouse) 244585, (Rat) 307724
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support