Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Peptide Sequence: MVRYSLDPED PTKSCKSRGS NLRVHFKNTR ETAQAIKGMH IRKATKYLKD VTLQKQCVPF RRYNGGVGRC AQAKQWGWTQ GRWPKKSAEF LLHMLKNAES NAELKGLDVD SLVIEHIQVN KAPKVRRRTY RAHGRINPYM SSPCHIEMIL TEKEQIVPKP EEEVAQKKKI SQKKLKKQKL MARE
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L22P family of ribosomal proteins. It is located in the cytoplasm. This gene has been referred to as rpL23 because the encoded protein shares amino acid identity with ribosomal protein L23 from Halobacterium marismortui; however, its official symbol is RPL17. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Alternative splicing results in multiple transcript variants. Read-through transcription also exists between this gene and the neighboring downstream C18orf32 gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 60S ribosomal protein L17; 60S ribosomal protein L23; gene encoding putative NFkB activating protein; Large ribosomal subunit protein uL22; MGC117162; PD-1
Gene Aliases: L17; PD-1; RPL17; RPL23
UniProt ID: (Human) P18621
Entrez Gene ID: (Human) 6139
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support