Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: MKFNPFVTS DRSKNRKRHF NAPSHIRRKI MSSPLSKELR QKYNVRSMPI RKDDEVQVVR GHYKGQQIGK VVQVYRKKYV IYIERVQREK ANGTTVHVGI HPSKVVITRL KLDKDRKKIL ERKAKSRQVG KEKGKYKEET IEKMQE (1-145 aa encoded by B C071664)
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L24P family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 60S ribosomal protein L26; Large ribosomal subunit protein uL24
Gene Aliases: DBA11; L26; RPL26
UniProt ID: (Human) P61254
Entrez Gene ID: (Human) 6154
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support