Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPRKIEEIK DFLLTARRKD AKSVKIKKNK DNVKFKVRCS RYLYTLVITD KEKAEKLKQS LPPGLAVKEL K (1-70 aa encoded by BC000603)
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L38E family of ribosomal proteins. It is located in the cytoplasm. Alternative splice variants have been identified, both encoding the same protein. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome, including one located in the promoter region of the type 1 angiotensin II receptor gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 60S ribosomal protein L38; Large ribosomal subunit protein eL38
Gene Aliases: 0610025G13Rik; L38; Rbt; RPL38; Ts; Tss
UniProt ID: (Human) P63173, (Rat) P63174, (Mouse) Q9JJI8
Entrez Gene ID: (Human) 6169, (Rat) 689284, (Mouse) 67671
Molecular Function:
ribosomal protein
translational protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support