Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GSASSEAPP LINEDVKRTV DLSSHLAKVT AEVVLAHLGG GSTSRATSFL LALEPELEAR LAHLGVQVKG EDEEENNLEV RETKIKGKSG RFFTVKLPVA LDPGAKISVI VETVYTHVLH PYPTQITQSE KQFVVFEGNH YFYSPYPTKT QTMRVKLASR NVESYTKLGN PTRSEDLLDY GPFRDVPAYS QDTFKVHYEN NSPFLTITSM TRVIEVSHWG NIAVEENVDL KHTGAVLKGP FSRYDYQRQP DSGISSIRSF KTILPAAAQD VYYRDEIGNV STSHLLILDD SVEMEIRPRF PLFGGWKTHY IVGYNLPSYE YLYNLGDQYA (22-350 aa encoded by BC010839)
Ribophorins 1 and 2 are abundant and highly conserved glycoproteins residing in the endoplasic reticulum, that participate in ribosome binding. Mammalian oligosaccharyltransferase activity is associated with a protein complex composed of RPN1, RPN2, and an oligosaccharyltransferase protein. RPN1 is a component of the proteasome base. The ubiquitin-like (UBL) domain of recombinant Rad23 interacts with proteasomes through the leucine-rich repeat domain of RPN1. The RPN1 gene maps to chromosome 3 in somatic cell hybrids, and the RPN2 gene maps to chromosome 20 by in situ hybridization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dolichyl-diphosphooligosaccharide--protein; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase 67 kDa subunit; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit 1; dolichyl-diphosphooligosaccharide-protein glycosyltransferase 67 kDa subunit; oligosaccharyltransfe; oligosaccharyltransferase 1 homolog; oligosaccharyltransferase complex subunit (non-catalytic); Ribophorin I; Ribophorin-1; RPN-I
Gene Aliases: AU018702; D6Wsu137e; OST1; RBPH1; RIBI; Rpn-1; RPN-I; RPN1
UniProt ID: (Human) P04843, (Mouse) Q91YQ5
Entrez Gene ID: (Human) 6184, (Rat) 25596, (Mouse) 103963
Molecular Function: glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support