Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LTKLRFLQTE DSWVPASPDT GLDPLTVRRH VPAVWVLLSR DPLDPNECGY QPPGAPPGLG SMPSSSCGPR SRRRARDTRS
Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%.
May be a component of ribonuclease P or MRP.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alba-like protein C9orf23; Ribonuclease P protein subunit p25-like protein; ribonuclease P/MRP 25kDa subunit-like; RNase P protein subunit-like p25; Rpp25-like protein
Gene Aliases: bA296L22.5; C9orf23; RPP25L
UniProt ID: (Human) Q8N5L8
Entrez Gene ID: (Human) 138716
Molecular Function:
endoribonuclease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support