Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: YNYRVSFLIP ECGILSEELK NLVMNTGPYY FVKNLPLHEL ITPEFISTFI KKGSCYALTY NTHIDEDNTV ALLPNGKL
Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 81%.
Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5'-ends.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ribonuclease P (40 kD); Ribonuclease P protein subunit p40; ribonuclease P, 40kD subunit; ribonuclease P/MRP 40kDa subunit; ribonuclease P1; RNase P subunit 1; RNaseP protein p40
Gene Aliases: bA428J1.3; RNASEP1; RPP40
UniProt ID: (Human) O75818
Entrez Gene ID: (Human) 10799
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support