Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody specifically recognizes the 15 kDa RPS12 protein.
Immunogen sequence: MAEEGIAAG GVMDVNTALQ EVLKTALIHD GLARGIREAA KALDKRQAHL CVLASNCDEP MYVKLVEALC AEHQINLIKV DDNKKLGEWV GLCKIDREGK PRKVVGCSCV VVKDYGKESQ AKDVIEEYFK CKK (1-132 aa encoded by BC017321)
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S12E family of ribosomal proteins. It is located in the cytoplasm. Increased expression of this gene in colorectal cancers compared to matched normal colonic mucosa has been observed. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 40S ribosomal protein S12; Small ribosomal subunit protein eS12
Gene Aliases: RPS12; S12
UniProt ID: (Human) Q76M58, (Mouse) P63323
Entrez Gene ID: (Human) 6206, (Rat) 65139, (Mouse) 20042
Molecular Function: ribosomal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support