- Order Status
- Quick Order
-
Don't have an account ? Create Account
15603-1-AP is a rabbit polyclonal antibody raised against full-length RPS16 of human origin.
Immunogen sequence: MPSKGPLQS VQVFGRKKTA TAVAHCKRGN GLIKVNGRPL EMIEPRTLQY KLLEPVLLLG KERFAGVDIR VRVKGGGHVA QIYAIRQSIS KALVAYYQKY VDEASKKEIK DILIQYDRTL LVADPRRCES KKFGGPGARA RYQKSYR (1-146 aa encoded by BC004324)
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S9P family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 40S ribosomal protein S16; Small ribosomal subunit protein uS9
Gene Aliases: AA420385; AI317031; RPS16; S16
UniProt ID: (Human) P62249, (Mouse) P14131
Entrez Gene ID: (Human) 6217, (Mouse) 20055
Molecular Function:
ribosomal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support