Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MSLVIPEKFQ HILRVLNTNI DGRRKIAFAI TAIKGVGRRY AHVVLRKADI DLTKRAGELT EDEVERVITI MQNPRQYKIP DWFLNRQKDV KDGKYSQVLA NGLDNKLRED LERLKKIRAH RGLRHFWGLR VRGQHTKTTG RRGRTVGVSK KK
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S13P family of ribosomal proteins. It is located in the cytoplasm. The gene product of the E. coli ortholog (ribosomal protein S13) is involved in the binding of fMet-tRNA, and thus, in the initiation of translation. This gene is an ortholog of mouse Ke3. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 40S ribosomal protein S18; Ke-3; Ke3; rhabdomyosarcoma antigen MU-RMS-40.21; Small ribosomal subunit protein uS13
Gene Aliases: D6S218E; HKE3; KE-3; KE3; RPS18; S18
UniProt ID: (Human) P62269
Entrez Gene ID: (Human) 6222
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support