Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GRGARGGKAE DKEWMPVTKL GRLVKDMKIK SLEEIYLFSL PIKESEIIDF FLGASLKDEV LKIMPVQKQT RAGQRTRFKA FVAIGDYNGH VGLGVKCSKE VATAIRGAII LAKLSIVPVR RGYW
Bacterial pathogens deliver type III effector proteins into the plant cell during infection. Plants express disease resistance (R) proteins that respond specifically to a particular type III effector by activating immune responses. Resistance proteins guard the plant against pathogens that contain an appropriate avirulence protein via an indirect interaction. RPS2 (resistance to Pseudomonas syringae protein 2) is a 909 amino acid plasma membrane protein with leucine-rich repeat, leucine zipper and P loop domains that confers resistance to Pseudomonas syringae infection by interacting with the avirulence gene AvrRpt2. Belonging to the disease resistance NB-LRR family, RPS2 directly associates with RIN4, which triggers plant resistance when RIN4 is degraded by AvrRpt2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 40S ribosomal protein S2; 40S ribosomal protein S4; OK/KNS-cl.6; Protein LLRep3; Small ribosomal subunit protein uS5
Gene Aliases: LLREP3; RPS2; RPS4; S2
UniProt ID: (Human) P15880
Entrez Gene ID: (Human) 6187
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support