Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAVSKNKRL TKGGKKGAKK KVVDPFSKKD WYDVKAPAMF NIRNIGKTLV TRTQGTKIAS DGLKGRVFEV SLADLQNDEV AFRKFKLITE DVQGKNCLTN FHGMDLTRDK MCSMVKKWQT MIEAHVDVKT TDGYLLRLFC VGFTKKRNNQ IRKTSYAQHQ QVRQIRKKMM EIMTREVQTN DLKEVVNKLI PDSIGKDIEK ACQSIYPLHD VFVRKVKMLK KPKFELGKLM ELHGEGSSSG KATGDETGAK VERADGYEPP VQESV (1-264 aa encoded by BC066926)
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S3AE family of ribosomal proteins. It is located in the cytoplasm. Disruption of the gene encoding rat ribosomal protein S3a, also named v-fos transformation effector protein, in v-fos-transformed rat cells results in reversion of the transformed phenotype. This gene is co-transcribed with the U73A and U73B small nucleolar RNA genes, which are located in its fourth and third introns, respectively. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Alternatively spliced transcript variants have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 40S ribosomal protein S3a; 40S ribosomal protein S3a {ECO:0000255|HAMAP-Rule:MF_03122}; Fte-1; Protein TU-11; ribosomal protein S3A; RPS3A; Small ribosomal subunit protein eS1; V-fos transformation effector protein; v-fos transformation effector protein 1
Gene Aliases: Fte-1; FTE1; MFTL; RPS3A; Rps3a1; S3A
UniProt ID: (Human) P61247, (Rat) P49242, (Mouse) P97351
Entrez Gene ID: (Human) 6189, (Rat) 29288, (Mouse) 20091
Molecular Function: ribosomal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support