Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTEWETAAPA VAETPDIKLF GKWSTDDVQI NDISLQDYIA VKEKYAKYLP HSAGRYAAKR FRKAQCPIVE RLTNSMMMHG RNNGKKLMTV RIVKHAFEII HLLTGENPLQ VLVNAIINSG PREDSTRIGR AGTVRRQAVD VSPLRRVNQA IWLLCTGARE AAFRNIKTIA ECLADELINA AKGSSNSYAI KKKDELERVA KSNR
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S7P family of ribosomal proteins. It is located in the cytoplasm. Variable expression of this gene in colorectal cancers compared to adjacent normal tissues has been observed, although no correlation between the level of expression and the severity of the disease has been found. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 40S ribosomal protein S5; Small ribosomal subunit protein uS7
Gene Aliases: RPS5; S5
UniProt ID: (Human) P46782
Entrez Gene ID: (Human) 6193
Molecular Function:
ribosomal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support