Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MPLAQLKEPW PLMELVPLDP ENGQTSGEEA GLQPSKDEGV LKEISITHHV KAGSEKADPS HFELLKVLGQ GSFGKVFLVR KVTRPDSGHL YAMKVLKKAT LKVRDRVRTK MERDILADVN HPFVVKLHYA FQTEGKLYLI LDFLRGGDLF TRLSKEVMFT EEDVKFYLAE LALGLDHLHS LGIIYRDLKP ENILLDEEGH IKLTDFGLSK EAIDHEKKAY SFCGTVEYMA PEVVNRQGHS HSADWWSYGV LMFEMLTGSL PFQGKDRKET MTLILKAKLG MPQFLSTEAQ SLLRALFKRN PANRLGSGPD GAEEIKRHVF YSTIDWNKLY RREIKPPFKP AVAQPDDTFY FDTEFTSRTP KDSPGIPPSA GAHQLFRGFS FVATGLMEDD GKPRAPQAPL HSVVQQLHGK NLVFSDGYVV KETIGVGSYS ECKRCVHKAT NMEYAVKVID KSKRDPSEEI EILLRYGQHP NIITLKDVYD DGKHVYLVTE LMRGGELLDK ILRQKFFSER EASFVLHTIG KTVEYLHSQG VVHRDLKPSN ILYVDESGNP ECLRICDFGF AKQLRAENGL LMTPCYTANF VAPEVLKRQG YDEGCDIWSL GILLYTMLAG YTPFANGPSD TPEEILTRIG SGKFTLSGGN WNTVSETAKD LVSKMLHVDP HQRLTAKQVL QHPWVTQKDK LPQSQLSHQD LQLVKGAMAA TYSALNSSKP TPQLKPIESS ILAQRRVRKL PSTTL
RPS6KA1 (RSK1) is a growth-factor regulated serine/threonine kinase that is involved in the MAPK cascade. It is thought to control cellular proliferation and differentiation through the phosphorylation of transcription factors. RPS6KA1 contains 2 nonidentical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 90 kDa ribosomal protein S6 kinase 1; dJ590P13.1 (ribosomal protein S6 kinase, 90kD, polypeptide 1); MAP kinase-activated protein kinase 1a; MAPK-activated protein kinase 1a; MAPKAP kinase 1a; MAPKAPK-1a; p90-RSK 1; p90RSK1; p90S6K; ribosomal protein S6 kinase alpha 1; Ribosomal protein S6 kinase alpha-1; ribosomal protein S6 kinase, 90kDa, polypeptide 1; Ribosomal S6 kinase 1; RSK-1; S6K-alpha 1; S6K-alpha-1
Gene Aliases: HU-1; MAPKAPK1A; p90Rsk; RPS6KA1; RSK; RSK1
UniProt ID: (Human) Q15418
Entrez Gene ID: (Human) 6195
Molecular Function:
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support