Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: HeLa.
Predicted reactivity: Mouse (96%), Rat (96%), Bovine (97%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RPS6KB2 (p70S6K beta) is a serine/threonine kinase with 2 non-identical kinase catalytic domains which phosphorylates the S6 ribosomal protein and eukaryotic translation initiation factor 4B (eIF4B). Phosphorylation of S6 leads to an increase in protein synthesis and cell proliferation.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 70 kDa ribosomal protein S6 kinase 2; p70 ribosomal S6 kinase beta; p70 S6 kinase beta; p70 S6K-beta; p70 S6KB; p70-S6K 2; P70S6K2; Ribosomal protein S6 kinase beta-2; ribosomal protein S6 kinase, 70kDa, polypeptide 2; S6 kinase-related kinase; S6K-beta; S6K-beta-2; serine/threonine-protein kinase 14 beta; Serine/threonine-protein kinase 14B; SRK
Gene Aliases: KLS; p70(S6K)-beta; P70-beta; P70-beta-1; P70-beta-2; p70S6Kb; RPS6KB2; S6K-beta2; S6K2; SRK; STK14B
UniProt ID: (Human) Q9UBS0
Entrez Gene ID: (Human) 6199
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support