Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VVDRKGPLVT LNKPQGLPVT GKPGELTLFS VLPELSQSLG LREQELQVVR ASGKESSGLV LLSSCPQTAS RLQKYFTHAR R
Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 60%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitochondrial mRNA pseudouridine synthase RPUSD3; RNA pseudouridylate synthase domain-containing protein 3
Gene Aliases: AI527266; RGD1307582; RPUSD3
UniProt ID: (Human) Q6P087, (Mouse) Q14AI6
Entrez Gene ID: (Human) 285367, (Rat) 362416, (Mouse) 101122
Molecular Function:
RNA processing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support