Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEESDSEKT TEKENLGPRM DPPLGEPEGS LGWVLPNTAM KKKVLLMGKS GSGKTSMRSI IFANYIARDT RRLGATIDVE HSHVRFLGNL VLNLWDCGGQ DTFMENYFTS QRDNIFRNVE VLIYVFDVES RELEKDMHYY QSCLEAILQN SPDAKIFCLV HKMDLVQEDQ RDLIFKEREE DLRRLSRPLE CSCFRTSIWD ETLYKAWSSI VYQLIPNVQQ LEMNLRNFAE IIEADEVLLF ERATFLVISH YQCKEQRDAH RFEKISNIIK QFKLSCSKLA ASFQSMEVRN SNFAAFIDIF TSNTYVMVVM SDPSIPSAAT LINIRNARKH FEKLERVDGP KQCLLMR (1-346 aa encoded by BC034726)
Ras-homologous GTPases constitute a large family of signal transducers that alternate between an activated, GTP-binding state and an inactivated, GDP-binding state. These proteins represent cellular switches that are operated by GTP-exchange factors and factors that stimulate their intrinsic GTPase activity. All GTPases of the Ras superfamily have in common the presence of six conserved motifs involved in GTP/GDP binding, three of which are phosphate-/magnesium-binding sites (PM1-PM3) and three of which are guanine nucleotide-binding sites (G1-G3). Transcript variants encoding distinct isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTP-binding protein ragB; Rag B; ragB; Ras-related GTP binding B; Ras-related GTP-binding protein B; RRAGB; rragB {ECO:0000312|RGD:619805}
Gene Aliases: bA465E19.1; RAGB; RRAGB
UniProt ID: (Human) Q5VZM2, (Rat) Q63487, (Mouse) Q6NTA4
Entrez Gene ID: (Human) 10325, (Rat) 117043, (Mouse) 245670
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support