Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSLQYGAEE TPLAGSYGAA DSFPKDFGYG VEEEEEEAAA AGGGVGAGAG GGCGP (1-54 aa encoded by BC016668)
RRAGC (Ras-related GTP-binding protein C) is a monomeric guanine nucleotide-binding protein that plays a crucial role in the cellular response to amino acid availability through regulations of the mTORC1 signaling cascade. RRAGC forms a heterodimer with RRAGA and RRAGB and is primarily localized to the cytoplasm. It cycles between an inactive GTP-bound and an active GDP-bound form. RRAGC, in its active form, participates in the re-localization of mTORC1 to the lysosomes and its subsequent activation by the GTPase RHEB. This re-localization is a crucial step in the activation of the TOR signaling cascade by amino acids.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTPase-interacting protein 2; Rag C; Rag C protein; Ras-related GTP binding C; Ras-related GTP-binding protein C; RRAGC; small GTPase homolog; small GTPase, homolog; TIB929
Gene Aliases: AU041672; GTR2; RAGC; RRAGC; TIB929; YGR163W
UniProt ID: (Human) Q9HB90, (Mouse) Q99K70
Entrez Gene ID: (Human) 64121, (Mouse) 54170
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support