Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
RRM2 is one of two non-identical subunits for ribonucleotide reductase. This reductase catalyzes the formation of deoxyribonucleotides from ribonucleotides. Synthesis of the protein (M2) is regulated in a cell-cycle dependent fashion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ribonucleoside-diphosphate reductase M2 chain; Ribonucleoside-diphosphate reductase subunit M2; Ribonucleotide reductase 2; ribonucleotide reductase M2 polypeptide; Ribonucleotide reductase small chain; Ribonucleotide reductase small subunit; RRR2M
Gene Aliases: AA407299; R2; RR2; RR2M; RRM2
UniProt ID: (Human) P31350, (Mouse) P11157, (Rat) Q4KLN6
Entrez Gene ID: (Human) 6241, (Mouse) 20135, (Rat) 362720
Molecular Function: reductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support