Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ETATQTCGG TDSTEGLFNM DEDEETEHET KAGPERLDQM VHPVAERLDI LMSLVLSYMK DVCYVDGKVD NGKTKDLYRD LINIFDKLLL PTHASCHVQ (268-365 aa encoded by BC132688)
Required for efficient transcription initiation by RNA polymerase I. Required for the formation of the competent preinitiation complex (PIC). Dissociates from pol I as a consequence of transcription. In vitro, cannot activate transcription in a subsequent transcription reaction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RNA polymerase I-specific transcription initiation factor RRN3; RRN3 RNA polymerase I transcription factor homolog; TIF-IA; Transcription initiation factor IA; transcription initiation factor TIF-IA
Gene Aliases: A-270G1.2; AL023001; E130302O19Rik; R75565; RRN3; TIF-1A; Tif1a; TIFIA
UniProt ID: (Human) Q9NYV6, (Mouse) B2RS91
Entrez Gene ID: (Human) 54700, (Mouse) 106298
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support