Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEGQSVEEL LAKAEQDEAE KLQRITVHKE LELQFDLGNL LASDRNPPTG LRCAGPTPEA ELQALARDNT QLLINQLWQL PTERVEEAIV ARLPEPTTRL PREKPLPRPR PLTRWQQFAR LKGIRPKKKT NLVWDEVSGQ WRRRWGYQRA RDDTKEWLIE VPGNADPLED QFAKRIQAKK ERVAKNELNR LRNLARAHKM QLPSAAGLHP TGHQSKEELG RAMQVAKVST ASVGRFQERL PKEKVPRGSG KKRKFQPLFG DFAAEKKNQL ELLRVMNSKK PQLDVTRATN KQMREEDQEE AAKRRKMSQK GKRKGGRQGP GGKRKGGPPS QGGKRKGGLG GKMNSGPPGL GGKRKGGQRP GGKRRK (1-365 aa encoded by B C001811)
Involved in ribosomal large subunit assembly. May regulate the localization of the 5S RNP/5S ribonucleoprotein particle to the nucleolus. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homolog of yeast ribosome biogenesis regulator; homolog of yeast ribosome biogenesis regulatory protein RRS1; regulator for ribosome resistance homolog; Ribosome biogenesis regulatory protein homolog; ribosome biogenesis regulatory protein RRS1 homolog; RRS1 ribosome biogenesis regulator homolog
Gene Aliases: 5730466A07Rik; AA415037; D1Ertd701e; KIAA0112; MNCb-2643; RRR; RRS1
UniProt ID: (Human) Q15050, (Mouse) Q9CYH6, (Rat) A1A5P2
Entrez Gene ID: (Human) 23212, (Mouse) 59014, (Rat) 297784
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support