Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
It is not recommended to aliquot this antibody.
This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 nonidentical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 90 kDa ribosomal protein S6 kinase 1; 90 kDa ribosomal protein S6 kinase 2; 90 kDa ribosomal protein S6 kinase 3; dJ590P13.1 (ribosomal protein S6 kinase, 90kD, polypeptide 1); Insulin-stimulated protein kinase 1; ISPK-1; MAP kinase-activated protein kinase 1; MAP kinase-activated protein kinase 1a; MAP kinase-activated protein kinase 1b; MAP kinase-activated protein kinase 1c; MAPK-activated protein kinase 1; MAPK-activated protein kinase 1a; MAPK-activated protein kinase 1b; MAPK-activated protein kinase 1c; MAPKAP kinase 1; MAPKAP kinase 1a; MAPKAP kinase 1b; MAPKAP kinase 1C; MAPKAPK-1; MAPKAPK-1a; MAPKAPK-1b; MAPKAPK-1c; MAPKAPK1A; mitogen-activated protein kinase-activated protein kinase 1C; p90 rsk; p90-RSK; p90-RSK 1; p90-RSK 2; p90-RSK 3; p90RSK1; p90RSK2; p90S6K; pp90RSK1; pp90RSK2; pp90RSK3; Protein-tyrosine kinase Mpk-9; ribosomal protein S6 kinase 2; Ribosomal protein S6 kinase 2 alpha; ribosomal protein S6 kinase alpha 1; ribosomal protein S6 kinase alpha 2; Ribosomal protein S6 kinase alpha-1; Ribosomal protein S6 kinase alpha-2; Ribosomal protein S6 kinase alpha-3; Ribosomal protein S6 kinase II alpha; ribosomal protein S6 kinase polypeptide 1; ribosomal protein S6 kinase, 90kD, polypeptide 1; Ribosomal protein S6 kinase, 90kD, 1; ribosomal protein S6 kinase, 90kD, polypeptide 2; ribosomal protein S6 kinase, 90kDa, polypeptide 1; ribosomal protein S6 kinase, 90kDa, polypeptide 2; ribosomal protein S6 kinase, 90kDa, polypeptide 3; Ribosomal S6 kinase 1; Ribosomal S6 kinase 2; Ribosomal S6 kinase 3; RP11-393H10.3; RSK-1; RSK-2; RSK-3; S6 protein kinase (Rsk-1); S6K-alpha 1; S6K-alpha 2; S6K-alpha-1; S6K-alpha-2; S6K-alpha-3; S6KII-alpha
Gene Aliases: 90kDa; C130006E23; CLS; D17Wsu134e; HU-1; HU-2; HU-3; ISPK-1; ISPK1; Mapkapk-1a; MAPKAPK-1b; MAPKAPK1A; MAPKAPK1B; MAPKAPK1C; MPK-9; MRX19; p90-Rsk1; p90-RSK2; p90-RSK3; p90Rsk; p90Rsk1; p90RSK2; p90RSK3; p90S6K; pp90rsk; pp90RSK2; pp90RSK3; RGD1563860; RPS6KA; Rps6ka-rs1; RPS6KA1; RPS6KA1L; RPS6KA2; RPS6KA3; RSK; Rsk-1; RSK1; RSK2; RSK3; S6K-alpha; S6K-alpha-1; S6K-alpha2; S6K-alpha3
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support