Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a protein that appears to be a component the radial spoke head, as determined by homology to similar proteins in the biflagellate alga Chlamydomonas reinhardtii and other ciliates. Radial spokes, which are regularly spaced along cilia, sperm, and flagella axonemes, consist of a thin 'stalk' and a bulbous 'head' that form a signal transduction scaffold between the central pair of microtubules and dynein. Mutations in this gene cause primary ciliary dyskinesia 1, a disease arising from dysmotility of motile cilia and sperm. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Radial spoke head protein 4 homolog A; Radial spoke head-like protein 3; radial spokehead-like 3
Gene Aliases: A230081C05; CILD11; dJ412I7.1; RSHL3; RSPH4A; RSPH6B
UniProt ID: (Human) Q5TD94, (Mouse) Q8BYM7
Entrez Gene ID: (Human) 345895, (Rat) 309767, (Mouse) 212892
Molecular Function:
microtubule or microtubule-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support