Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: DQNVKFRKLM GIKSEDEAGC SSVDEESYKT LKQQEEVFRN LDAQYEMARS
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%
In vitro study revealed that RSRC2 might play a role in cell proliferation. RSRC2 may be a novel tumor suppressor of esophageal cancer cell growth.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: arginine/serine-rich coiled-coil 2; Arginine/serine-rich coiled-coil protein 2; FLJ11021
Gene Aliases: RSRC2
UniProt ID: (Human) Q7L4I2
Entrez Gene ID: (Human) 65117
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support