Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RPWRLRCPA LHLPSLSVFS LRWKLPSLTT DETMCKSVTT DEWKKVFYEK MEEAKPADSW DLIIDPNLKH NVLSPGWKQY LELHASGRFH CSWCWHTWQS PYVVILFHMF LDRAQRAGSV RMRVFKQLCY ECGTARLDES SMLEENIEGL VDNLITSLRE QCYGERGGQY RIHVASRQDN RRHRGEFCEA CQEGIVHWKP SEKLLEEEAT TYTFSRAPSP TKSQDQ (3-227 aa encoded by B C034744)
Specifically promotes functional cell surface expression of olfactory receptors, but not of other GPCRs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3CxxC-type zinc finger protein 1; receptor (chemosensory) transporter protein 1; receptor transporter protein 1; receptor transporting protein 1; Receptor-transporting protein 1; zinc finger, 3CxxC-type 1
Gene Aliases: Gm1766; Gm604; RGD1560980; RTP1; Z3CXXC1
UniProt ID: (Human) P59025, (Mouse) Q8C8C1
Entrez Gene ID: (Human) 132112, (Mouse) 239766, (Rat) 288006
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support