Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TKIDGLEKT NSKLQEELSA ATDRICSLQE EQQQLREQNE LIRERSEKSV EITKQDTKVE LETYKQTRQG LDEMYSDVWK QLKEEKKVRL ELEKELELQI GMKTEMEIAM KLLEKDTHEK QDTLVALRQQ LEEVKAINLQ MFHKAQNAES SLQQKNEAIT SFEGKTNQVM SSMKQMEERL QHSERARQGA EERSHKLQQE LGGRIGALQL QLSQLHEQCS SLEKELKSEK EQRQALQREL QHEKDTSSLL RMELQQVEGL KKELRELQDE KAELQKICEE QEQALQEMGL HLSQSKLKME DIKEVNQALK GHAWLKDDEA THCRQCEKEF SISRRKHHCR NCGHIFCNTC SSNELALPSY PKPVRVCDSC HTLLLQRCSS TAS (325-708 aa encoded by BC032571)
This gene encodes a protein that contains a RUN domain and a FYVE-type zinc finger domain. The encoded protein binds to phosphatidylinositol-3-phosphate (PI3P) and plays a role in early endosomal trafficking, tethering and fusion through interactions with small GTPases including Rab4, Rab5 and Rab14. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FYVE-finger containing protein; FYVE-finger protein EIP1; La-binding protein 1; Rab4-interacting protein; Rab4a interacting protein; RUN and FYVE domain-containing protein 1; Zinc finger FYVE domain-containing protein 12
Gene Aliases: 3000002E04Rik; RABIP4; RUFY1; ZFYVE12
UniProt ID: (Human) Q96T51, (Mouse) Q8BKQ4
Entrez Gene ID: (Human) 80230, (Mouse) 216724, (Rat) 360521
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support