Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: MATKDPTAV ERANLLNMAK LSIKGLIESA LSFGRTLDSD YPPLQQFFVV MEHCLKHGLK VRKSFLSYNK TIWGPLELVE KLYPEAEEIG ASVRDLPGLN EFYEYHALMM EEEGAVIVGL LVGLNVIDAN LCVKGEDLDS QVGVIDFSMY LKNEEDIGNK ERNVQIAAIL DQKNYVEELN RQLNSTVSSL HSRVDSLEKS NTKLIEELAI AKNNIIKLQE ENHQLRSENK LILMKTQQHL EVTKVDVETE LQTYKHSRQG LDEMYNEARR QLRDESQLRQ DVENELAVQV SMKHEIELAM KLLEKDIHEK QDTLIGLRQQ LEEVKAINIE MYQKLQGSED GLKEKNEIIA RLEEKTNKIT AAMRQLEQSD NDLLTQTRTI AMSLVKCASS DTQDQYKLVK DISF (1-403 aa encoded by BC041092)
RUFY2 (RUN And FYVE Domain Containing 2) is a Protein Coding gene. An important paralog of this gene is RUNDC3B.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: antigen MU-RMS-40.17; Leucine zipper FYVE-finger protein; LZ-FYVE; Rab4-interacting protein related; RUN and FYVE domain-containing 2; RUN and FYVE domain-containing protein 2; Run- and FYVE-domain containing protein
Gene Aliases: 2610111M19Rik; AI844453; Denn; KIAA1537; LZ-FYVE; RABIP4R; RUFY2; ZFYVE13
UniProt ID: (Human) Q8WXA3, (Mouse) Q8R4C2
Entrez Gene ID: (Human) 55680, (Rat) 690777, (Mouse) 70432
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support