Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
RUFY2 (RUN And FYVE Domain Containing 2) is a Protein Coding gene. An important paralog of this gene is RUNDC3B.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: antigen MU-RMS-40.17; Leucine zipper FYVE-finger protein; LZ-FYVE; Rab4-interacting protein related; RUN and FYVE domain-containing 2; RUN and FYVE domain-containing protein 2; Run- and FYVE-domain containing protein
Gene Aliases: 2610111M19Rik; AI844453; Denn; KIAA1537; LZ-FYVE; RABIP4R; RUFY2; ZFYVE13
UniProt ID: (Human) Q8WXA3, (Mouse) Q8R4C2
Entrez Gene ID: (Human) 55680, (Mouse) 70432
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support