Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
RUNX2, a nuclear protein, is a member of the RUNX family of transcription factors and has an Runt DNA-binding domain. It is essential for membranous and endochondral bone formation. It not only regulates osteoblastic differentiation and skeletal morphogenesis but also acts as a scaffold for nucleic acids and regulatory factors involved in skeletal gene expression. The protein can bind DNA both as a monomer and with more affinity, as a subunit of a heterodimeric complex. It plays a critical role in increasing TGFBR1 expression by osteoblasts and functions in cooperation with DLX5 or a related factor to activate osteoblast-specific gene expression. Mutations lead to disorders in bone development like cleidocranial dysplasia (CCD).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acute myeloid leukemia 3 protein; AKV core binding factor; AML-3; CBF-alpha-1; core binding factor alpha 1; Core-binding factor subunit alpha-1; core-binding factor, runt domain, alpha subunit 1; Oncogene AML-3; OSF-2; Osteoblast-specific transcription factor 2; PEA2-alpha A; PEBP2 alpha A; PEBP2-alpha A; Polyomavirus enhancer-binding protein 2 alpha A subunit; runt domain, alpha subunit 1; runt related transcription factor 2; Runt-related transcription factor 2; RUNX 2; RUNX-2; SL3-3 enhancer factor 1 alpha A subunit; SL3/AKV core-binding factor alpha A subunit
Gene Aliases: AML3; Cbf; CBF-alpha-1; Cbfa-1; CBFA1; CCD; CCD1; CLCD; LS3; OSF-2; OSF2; PEA2aA; PEBP2A; Pebp2a1; PEBP2aA; Pebpa2a; RUNX2
UniProt ID: (Human) Q13950, (Mouse) Q08775
Entrez Gene ID: (Human) 860, (Rat) 367218, (Mouse) 12393
Molecular Function:
DNA-binding transcription factor
immunoglobulin fold transcription factor
Runt transcription factor
gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support