Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PA1-32805 detects GPCR GPR100 from human samples.
GPR100 is a member of the rhodopsin family of G protein-coupled receptors (GPRs) and acts as a high affinity receptor for INSL5. Also acts as receptor for RLN3/relaxin-3, as well as bradykinin and kallidin. Binding of the ligand inhibits cAMP accumulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor 100; G-protein coupled receptor 100; G-protein coupled receptor GPCR142; Insulin-like peptide INSL5 receptor; JEG62; MGC126556; MGC126558; relaxin 3 receptor 2; Relaxin family peptide receptor 4; Relaxin-3 receptor 2; relaxin/insulin-like family peptide receptor 4; RLN3 receptor 2
Gene Aliases: GPCR142; GPR100; RLN3R2; RXFP4; RXFPR4
UniProt ID: (Human) Q8TDU9
Entrez Gene ID: (Human) 339403
Molecular Function: G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support