Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Retinoic Acid (RA; active metabolite of vitamin A) plays a prominent role in regulating the transition of proliferating precursor cells (such as carcinoma cells and neuronal precursors) to postmitotic differentiated cells (Joshi et al., 2005). The Retinoid X Receptors (RXRs) family (RXR alpha, beta, and gamma), preferentially bind 9-cis -RA and regulate gene transcription by forming heterodimers with a second family of RA receptors. RAs have been suggested to potentially play a therapeutic role in cervical cancer (Abu et al., 2005). RAs are known to play key roles in neuronal development and an increasing body of evidence indicates that retinoid signaling may regulate synaptic plasticity and associated learning and memory behaviors (Lane and Bailey, 2005).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Nuclear receptor subfamily 2 group B member 3; Retinoic acid receptor RXR-gamma; retinoic X receptor gamma-1; retinoid X receptor; Retinoid X receptor gamma; retinoid X receptor, gamma; RXR gamma; RXR gamma-1
Gene Aliases: NR2B3; RXRC; RXRG
UniProt ID: (Human) P48443, (Rat) Q5BJR8, (Mouse) P28705
Entrez Gene ID: (Human) 6258, (Rat) 83574, (Mouse) 20183
Molecular Function:
C4 zinc finger nuclear receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support