Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody can recognize the RXR beta protein and detects a 58 kDa band.
Immunogen sequence: RDGMGDSGR DSRSPDSSSP NPLPQGVPPP SPPGPPLPPS TAPSLGGSGA PPPPPMPPPP LGSPFPVISS SMGSPGLPPP APPGFSGPVS SPQINSTVSL PGGGSGPPED VKPPVLGVRG LHCPPPPGGP GAGKRLCAIC GDRSSGKHYG VYSCEGCKGF FKRTIRKDLT (70-238 aa encoded by BC001167)
This gene encodes a member of the retinoid X receptor (RXR) family of nuclear receptors which are involved in mediating the effects of retinoic acid (RA). This receptor forms homodimers with the retinoic acid, thyroid hormone, and vitamin D receptors, increasing both DNA binding and transcriptional function on their respective response elements. The gene lies within the major histocompatibility complex (MHC) class II region on chromosome 6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DADB-100D22.1; MGC1831; MHC class I promoter binding protein; MHC class I regulatory element-binding protein H-2RIIBP; nuclear receptor co-regulator 1; nuclear receptor coregulator 1; Nuclear receptor subfamily 2 group B member 2; putative retinoid X receptor beta; Retinoic acid receptor RXR-beta; retinoic acid receptor, beta; retinoid X receptor; Retinoid X receptor beta; retinoid X receptor, beta; RXR Beta
Gene Aliases: AL023085; DAUDI6; H-2RIIBP; NR2B2; RCoR-1; Rub; RXR-beta; RXRB
UniProt ID: (Human) P28702, (Mouse) P28704
Entrez Gene ID: (Human) 6257, (Rat) 361801, (Mouse) 20182
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support