Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is specific for the ~48 kDa RXR-beta protein.
This gene encodes a member of the retinoid X receptor (RXR) family of nuclear receptors which are involved in mediating the effects of retinoic acid (RA). This receptor forms homodimers with the retinoic acid, thyroid hormone, and vitamin D receptors, increasing both DNA binding and transcriptional function on their respective response elements. The gene lies within the major histocompatibility complex (MHC) class II region on chromosome 6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DADB-100D22.1; MGC1831; MHC class I regulatory element-binding protein H-2RIIBP; nuclear receptor co-regulator 1; nuclear receptor coregulator 1; Nuclear receptor subfamily 2 group B member 2; putative retinoid X receptor beta; Retinoic acid receptor RXR-beta; retinoic acid receptor, beta; retinoid X receptor; Retinoid X receptor beta; RXR Beta
Gene Aliases: AL023085; H-2RIIBP; Nr2b2; RCoR-1; Rub; RXR-beta; Rxrb
UniProt ID: (Mouse) P28704
Entrez Gene ID: (Rat) 361801, (Mouse) 20182
Molecular Function:
C4 zinc finger nuclear receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support