Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Secondary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Host
            Target Species
            Conjugate
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Secondary Antibodies ›
            • Rabbit anti-Goat IgG Secondary Antibodies

            Invitrogen

            Rabbit anti-Goat IgG (H+L) Secondary Antibody, HRP

            22 Published Figures
            212 References
            View all (67) Rabbit anti-Goat IgG antibodies
            Datasheet
            Create Citation
            Questions & Answers
            Catalog # 31402
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (KRW)
            280,000
            Online offer
            Ends: 30-Jun-2025
            329,000
            Save 49,000 (15%)
            1.5 mL
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Create Citation
            Questions & Answers

            Cite Rabbit anti-Goat IgG (H+L) Secondary Antibody, HRP

            • Antibody Testing Data (2)
            • Published Figures (22)
            Application
            Goat IgG (H+L) Secondary Antibody in Western Blot (WB)
            Group 53 Created with Sketch.

            FIGURE: 1 / 24

            Goat IgG (H+L) Secondary Antibody (31402) in WB

            Western blot was performed using Rabbit anti-Goat IgG (H+L) Secondary Antibody, HRP (Product # 31402). Whole cell extracts of LnCaP (Lane 1, 2, 3, 4, 5), HeLa (Lane 6) and HaCaT (Lane 7) were electrophoresed using NuPAGE™ 4-12% Bis-Tris Protein Gel (Product # NP03222BOX). Resolved proteins were transferred onto a nitrocellulose membrane (Product # IB23001) by iBlot® 2 Dry Blotting System (Product # IB21001)... View More
            View Product

            Goat IgG (H+L) Secondary Antibody in Western Blot (WB)
            Goat IgG (H+L) Secondary Antibody in Western Blot (WB)
            Goat IgG (H+L) Secondary Antibody in Western Blot (WB)
            Goat IgG (H+L) Secondary Antibody in Western Blot (WB)
            Goat IgG (H+L) Secondary Antibody in Western Blot (WB)
            Goat IgG (H+L) Secondary Antibody in Western Blot (WB)
            Goat IgG (H+L) Secondary Antibody in Western Blot (WB)
            Goat IgG (H+L) Secondary Antibody in Western Blot (WB)
            Goat IgG (H+L) Secondary Antibody in Western Blot (WB)
            Goat IgG (H+L) Secondary Antibody in Western Blot (WB)
            Goat IgG (H+L) Secondary Antibody in Western Blot (WB)
            Goat IgG (H+L) Secondary Antibody in Western Blot (WB)
            Goat IgG (H+L) Secondary Antibody in Western Blot (WB)
            Goat IgG (H+L) Secondary Antibody in Western Blot (WB)
            Goat IgG (H+L) Secondary Antibody in Western Blot (WB)
            Goat IgG (H+L) Secondary Antibody in Western Blot (WB)
            Goat IgG (H+L) Secondary Antibody in Western Blot (WB)
            Goat IgG (H+L) Secondary Antibody in Western Blot (WB)
            Goat IgG (H+L) Secondary Antibody in Western Blot (WB)
            Goat IgG (H+L) Secondary Antibody in Western Blot (WB)
            Goat IgG (H+L) Secondary Antibody in Western Blot (WB)
            Goat IgG (H+L) Secondary Antibody in Western Blot (WB)
            Goat IgG (H+L) Secondary Antibody in Western Blot (WB)
            Goat IgG (H+L) Secondary Antibody in Western Blot (WB)

            Product Details

            31402

            Applications
            Tested Dilution

            Western Blot (WB)

            1:10,000-1:200,000

            Immunohistochemistry (IHC)

            1:500-1:5,000

            Immunocytochemistry (ICC/IF)

            1:500-1:5,000
            Product Specifications

            Species Reactivity

            Goat

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Secondary Antibody

            Conjugate

            HRP
            • AP
            • Biotin 
            • FITC
            • TRITC
            • Request custom conjugation

            Form

            Lyophilized

            Concentration

            0.8 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.6, with 15mg/mL BSA

            Contains

            no preservative

            Storage conditions

            4°C

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_228395

            Target

            IgG

            Antibody Form

            Whole Antibody

            Product Specific Information

            Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration.

            This antibody has been successfully used in Western blot, and ICC applications.

            Antibody Specificity: The antibody reacts with the heavy chains of goat IgG and with the light chains common to most goat immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species.

            Restoration and Storage: Store product at 4°C until opened. Restore with 1.5 mL distilled water (0.8 mg/mL after restoration). Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. To judge clarity, draw product into a pasteur pipette. Product may be stored for several weeks at 4°C as an undiluted liquid. After dilution, do not use for more than one day.

            To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C.

            Country of Origin: USA

            Target Information

            Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            korea | 대한민국 flag icon
            Korea | 대한민국

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-x6nlx:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0