Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
Positive Samples: HeLa, HepG2, Jurkat, C6, Mouse lung, Mouse kidney, Rat lung, Rat liver
Immunogen sequence: RLKQIEEQTI KAQKELEEQT RKALELDQER KRAKEEAERL EKERRAAEEA KSAIAKQAAD QMKNQEQLAA ELAEFTAKIA LLEEAKKKKE EEATEWQHKA F
Radixin is a cytoskeletal protein that may be important in linking actin to the plasma membrane. It is highly similar in sequence to both ezrin and moesin. The radixin gene has been localized by fluorescence in situ hybridization to 11q23. A truncated version representing a pseudogene (RDXP2) was assigned to Xp21.3. Another pseudogene that seemed to lack introns (RDXP1) was mapped to 11p by Southern and PCR analyses. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, May 2012]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ESP10; Radixin; radixin isoform
Gene Aliases: AA516625; DFNB24; RDX
UniProt ID: (Human) P35241, (Mouse) P26043
Entrez Gene ID: (Human) 5962, (Rat) 315655, (Mouse) 19684
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support