Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive control: Ras full length protein.
Cellular location: Cell membrane, Golgi apparatus membrane.
This gene belongs to the Ras oncogene family, whose members are related to the transforming genes of mammalian sarcoma retroviruses. The products encoded by these genes function in signal transduction pathways. These proteins can bind GTP and GDP, and they have intrinsic GTPase activity. This protein undergoes a continuous cycle of de- and re-palmitoylation, which regulates its rapid exchange between the plasma membrane and the Golgi apparatus. Mutations in this gene cause Costello syndrome, a disease characterized by increased growth at the prenatal stage, growth deficiency at the postnatal stage, predisposition to tumor formation, mental retardation, skin and musculoskeletal abnormalities, distinctive facial appearance and cardiovascular abnormalities. Defects in this gene are implicated in a variety of cancers, including bladder cancer, follicular thyroid cancer, and oral squamous cell carcinoma. Multiple transcript variants, which encode different isoforms, have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-H-ras; c-has/bas p21 protein; c-K-ras; c-Ki-ras; c-Kirsten-ras protein; c-ras-Ki-2 activated oncogene; cellular c-Ki-ras2; cellular c-Ki-ras2 proto-oncogene; GTP- and GDP-binding peptide B; GTPase HRas; GTPase Hras1; GTPase KRas; GTPase NRas; H-Ras-1; Ha-Ras; Ha-Ras1 proto-oncoprotein; Harvey rat sarcoma viral oncogene homolog; Harvey rat sarcoma viral oncoprotein; K RAS; K-Ras 2; K-ras p21 protein; Ki-Ras; Kirsten rat sarcoma viral oncogene homolog; N-ras protein part 4; Neuroblastoma RAS; neuroblastoma RAS viral (v-ras) oncogene homolog; oncogene KRAS2; p19 H-RasIDX protein; p21ras; PR310 c-K-ras oncogene; Ras family small GTP binding protein H-Ras; RASK; RASN; transformation gene: oncogene HAMSV; Transforming protein N-Ras; Transforming protein p21; v-Ha-ras Harvey rat sarcoma viral oncogene homolog; v-Ki-ras2 Kirsten rat sarcoma 2 viral oncogene homolog; v-ras neuroblastoma RAS viral oncogene homolog
Gene Aliases: ALPS4; C-BAS/HAS; C-H-RAS; C-HA-RAS1; C-K-RAS; CFC2; CMNS; CTLO; H-RASIDX; HAMSV; HRAS; HRAS1; K-RAS2A; K-RAS2B; K-RAS4A; K-RAS4B; KI-RAS; KRAS; KRAS1; KRAS2; N-ras; NCMS; NRAS; NRAS1; NS; NS3; NS6; p21ras; RALD; RASH1; RASK2
UniProt ID: (Human) Q9UCE2, (Human) P01116, (Human) P01111
Entrez Gene ID: (Human) 3265, (Human) 3845, (Human) 4893
Molecular Function:
small GTPase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support