Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
FIGURE: 1 / 3
AA Sequence of recombinant protein: SSKTLCSMEE AINERIQEVA GSLIFRAISS IGLECQSVTS RGDLATCPRG FAVTGCTCGS ACGSWDVRAE TTCHCQCAGM DWTGARCCRV QP.
Preparation: Produced in BALB/c mice immunized with highly pure Recombinant Human Resistin. Anti-Human Resistin-specific antibody was purified from cell culture fluid by Protein A affinity chromatography.
Sandwich ELISA: Assuming 100 µL/well, a concentration of 0.5-1.0µg/mL of this antibody will detect at least 5.0 pg/mL of Recombinant Human Resistin when used with PeproTech Biotinylated Antigen Affinity Purified Anti-Human Resistin (500-P183GBT) as the detection antibody at a concentration of approximately 1.25-2.5µg/mL.
Western Blot: To detect Human Resistin by Western Blot analysis this antibody can be used at a concentration of 0.5- 1.0µg/mL. When used in conjunction with compatible secondary reagents the detection limit for Recombinant Human Resistin is 0.1-0.2 ng/lane, under non-reducing conditions.
Adipose tissue is an energy reserve in animals and is strictly regulated. Adipose cells produce and secrete numerous physiologically important proteins, such as lipoprotein lipase (LPL), leptin, adipocyte complement related protein of 30 kDa (Acrp30), perilipin, and resistin. Resistin, also known as Adipocyte Secreted Factor (ADSF) and FIZZ3, is a cytokine that is specifically secreted by adipocytes. While the function of resistin is not well understood, it appears to have key paracrine roles and has a suggested role in type 2 diabetes. Studies show that in obese, diabetic mice the levels of serum resistin are increased, which has an antagonistic effect on insulin. Additional studies have shown that resistin causes insulin resistance and glucose intolerance in mice. Resistin gene expression requires C/EBP alpha, which is necessary and sufficient for expression. Studies show that the coactivators CBP and p300, which have been shown to display histone acetyltransferase (HAT) activity, abundantly acetylate the histones at the resistin promoter suggesting their role in activating chromatin for resistin transcription. Numerous PPAR gamma and RXR ligands have been shown to downregulate resistin expression in adipocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adipose tissue specific secretory factor; Adipose tissue-specific secretory factor; ADSF; C/EBP epsilon regulated myeloid specific secreted cysteine rich protein precursor 1; C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich protein precursor 1; C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein; cysteine-r; Cysteine-rich secreted protein A12-alpha-like 2; Cysteine-rich secreted protein FIZZ3; dri; found in inflammatory zone 3; MGC126603; MGC126609; Resistin; resistin delta2; RSN
Gene Aliases: ADSF; FIZZ3; HXCP1; RETN; RETN1; RSTN; UNQ407/PRO1199; XCP1
UniProt ID: (Human) Q9HD89
Entrez Gene ID: (Human) 56729
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support