Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Invitrogen
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
RhoA is described as a small GTPase protein that regulates the actin cytoskeleton in the formation of stress fibers. RhoA is also included in the related proteins of the Ras superfamily, which involves proteins in the regulation and timing of cell division.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aplysia ras-related homolog 12; Aplysia RAS-related homolog 6; aplysia ras-related homolog 9 (RhoC); aplysia ras-related homolog A; aplysia ras-related homolog A1; aplysia ras-related homolog A2; aplysia ras-related homolog B; h12; h6; h9; oncogene RHO H12; oncogene RHO H6; oncogene RHO H9; plysia ras-related homolog A2; Ras family member A; ras homolog 9 (RhoC); ras homolog A1; ras homolog A2; ras homolog B; ras homolog gene family, member A; ras homolog gene family, member A1; ras homolog gene family, member A2; ras homolog gene family, member AB; ras homolog gene family, member B; ras homolog gene family, member C; RAS-related homolog 9; Rho; Rho cDNA clone 12; Rho cDNA clone 6; Rho cDNA clone 9; Rho family GTPase; Rho-related GTP-binding protein RhoB; Rho-related GTP-binding protein RhoC; rhoB; rhoC GTPase; SIG-61; Silica-induced gene 61 protein; small GTP binding protein RhoA; small GTP binding protein RhoC; Transforming protein RhoA
Gene Aliases: AA017882; AI324259; ARH12; ARH6; ARH9; ARHA; Arha1; Arha2; ARHB; ARHC; H9; MST081; MSTP081; RHO12; RHOA; RHOB; RHOC; RHOH12; RHOH6; RHOH9
UniProt ID: (Human) P61586, (Human) P62745, (Human) P08134, (Rat) P61589, (Mouse) Q9QUI0, (Rat) P62747, (Mouse) P62746, (Mouse) Q62159
Entrez Gene ID: (Human) 387, (Human) 388, (Human) 389, (Rat) 117273, (Mouse) 11848, (Rat) 64373, (Mouse) 11852, (Mouse) 11853, (Rat) 295342
Molecular Function:
small GTPase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support