Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Positive Control: U87-MG, mouse brain, rat brain
Predicted Reactivity: Xenopus laevis (90%), Dog (99%), Pig (100%), Chicken (97%), Sheep (100%), Rhesus Monkey (89%), Bovine (100%)
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Rho-related GTP-binding protein (RhoB) mediates apoptosis in neoplastically transformed cells after DNA damage. While not essential for development, it does affect cell adhesion and growth factor signaling in transformed cells. RhoB targets PKN1 to endosomes and is involved in trafficking of the EGF receptor from late endosomes to lysosomes. Also required for stability and nuclear trafficking of AKT1/AKT which promotes endothelial cell survival during vascular development. Serves as a tumor supressor - deletion of RhoB causes tumor formation.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aplysia RAS-related homolog 6; aplysia ras-related homolog B; h6; oncogene RHO H6; ras homolog B; ras homolog gene family, member AB; ras homolog gene family, member B; Rho; Rho cDNA clone 6; Rho-related GTP-binding protein RhoB; rhoB
Gene Aliases: AA017882; ARH6; ARHB; MST081; MSTP081; RHOB; RHOH6
UniProt ID: (Human) P62745, (Rat) P62747, (Mouse) P62746
Entrez Gene ID: (Human) 388, (Rat) 64373, (Mouse) 11852
Molecular Function: small GTPase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support