Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Aspire Free Products
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • RhoB Antibodies
            Formerly called “Recombinant polyclonal antibody”, this product is now rebranded as “Recombinant Superclonal™ antibody”. The physical product and the performance remain unchanged.

            Invitrogen

            RhoB Recombinant Superclonal™ Antibody (19HCLC)

            Advanced Verification
            2 References
            View all (16) RhoB antibodies
            Datasheet
            Protocols
            Questions & Answers
            Tech Support
            Catalog # 711274
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (EUR)
            514,00
            100 µg
             
            Estimated availability date: 19-Aug-2025
            Check your price
            Request bulk or custom format
            Datasheet
            Protocols
            Questions & Answers
            Tech Support

            Cite RhoB Recombinant Superclonal™ Antibody (19HCLC)

            • Antibody Testing Data (2)
            • Advanced Verification (1)
            Application
            RhoB Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 3

            RhoB Antibody (711274) in ICC/IF

            For immunofluorescence analysis, UV (302 nm, 60 minutes) treated A549 cells were fixed and permeabilized for detection of endogenous RhoB using Anti-RhoB Recombinant Rabbit Superclonal™ Antibody (Product # 711274, 2 µg/mL) and labeled with Goat anti-Rabbit IgG (Heavy Chain) Superclonal™ Secondary Antibody, Alexa Fluor® 488 conjugate (Product # A27034, 1:2000). Panel a) shows representative cells that were stained for detection and localization of RhoB protein (green), Panel b) is stained for nuclei (blue) using SlowFade® Gold Antifade Moun... View More
            View Product

            RhoB Antibody in Immunocytochemistry (ICC/IF)
            RhoB Antibody in Western Blot (WB)
            RhoB Antibody

            Product Details

            711274

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1-2 µg/mL
            View 2 publications 2 publications

            Immunocytochemistry (ICC/IF)

            2 µg/mL
            -
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Published species

            Human

            Host/Isotype

            Rabbit / IgG

            Expression System

            Expi293

            Class

            Recombinant Superclonal

            Type

            Antibody

            Clone

            19HCLC

            Immunogen

            Peptides corresponding to Human RHOB (aa 128-145, 50-65)
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.5 mg/mL

            Purification

            Protein A

            Storage buffer

            PBS, pH 7.2

            Contains

            0.09% sodium azide

            Storage conditions

            Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

            Shipping conditions

            Wet ice

            RRID

            AB_2633147

            Product Specific Information

            This antibody is predicted to react with Mouse, Monkey, Goat and Cat.

            Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called “Recombinant polyclonal antibody”, this product is now rebranded as “Recombinant Superclonal™ antibody”. The physical product and the performance remain unchanged.

            Target Information

            Rho-related GTP-binding protein (RhoB) mediates apoptosis in neoplastically transformed cells after DNA damage. While not essential for development, it does affect cell adhesion and growth factor signaling in transformed cells. RhoB targets PKN1 to endosomes and is involved in trafficking of the EGF receptor from late endosomes to lysosomes. Also required for stability and nuclear trafficking of AKT1/AKT which promotes endothelial cell survival during vascular development. Serves as a tumor supressor - deletion of RhoB causes tumor formation.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Related Products for

            Bioinformatics

            Protein Aliases: Aplysia RAS-related homolog 6; aplysia ras-related homolog B; h6; oncogene RHO H6; ras homolog B; ras homolog gene family, member AB; ras homolog gene family, member B; Rho; Rho cDNA clone 6; Rho-related GTP-binding protein RhoB; rhoB

            View more View less

            Gene Aliases: AA017882; ARH6; ARHB; MST081; MSTP081; RHOB; RHOH6

            View more View less

            UniProt ID: (Human) P62745, (Rat) P62747, (Mouse) P62746

            View more View less

            Entrez Gene ID: (Human) 388, (Rat) 64373, (Mouse) 11852

            View more View less

            Function(s)

            GTPase activity protein binding GTP binding GDP binding nucleotide binding small GTPase

            Process(es)

            cytokinesis angiogenesis intracellular protein transport apoptotic process cell adhesion Rho protein signal transduction endosome to lysosome transport positive regulation of endothelial cell migration cell differentiation platelet activation regulation of cell migration negative regulation of cell migration positive regulation of apoptotic process positive regulation of angiogenesis negative regulation of cell cycle regulation of small GTPase mediated signal transduction endothelial tube morphogenesis cellular response to hydrogen peroxide cellular response to ionizing radiation obsolete transformed cell apoptotic process small GTPase mediated signal transduction transport multicellular organism development protein transport
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            • Price & Freight Policy
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • German Supply Chain Due Diligence Act
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Legal Notice
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            germany | deutschland flag icon
            Germany | Deutschland

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-mgmkt:80/100.66.75.22:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0