Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Store vial at 4° C prior to restoration. For extended storage aliquot contents and freeze at -20° C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
Anti-RIBONUCLEASE A is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum as well as purified and partially purified Ribonuclease A [Bovine pancreas]. Cross reactivity against Ribonuclease A from other tissues and species may occur but have not been specifically determined.
Endonuclease that catalyzes the cleavage of RNA on the 3' side of pyrimidine nucleotides. Acts on single stranded and double stranded RNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Rib 1; ribonuclease A (pancreatic); ribonuclease A C1; Ribonuclease BS-1; Ribonuclease pancreatic; RNase 1; RNase A; RNAseA; S-RNase; Seminal ribonuclease; seminal RNase
Gene Aliases: RAC1; RNASE1; RNS1; SRN
UniProt ID: (Bovine) P00669, (Bovine) P61823
Entrez Gene ID: (Bovine) 280930, (Bovine) 282340
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support