Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Immunogen sequence: APGSASSEAP PLINEDVKRT VDLSSHLAKV TAEVVLAHLG GGSTSRATSF LLALEPELEA RLAHLGVQVK GEDEEENNLE VRETKIKGKS GRFFTVKLPV ALDPGAKISV IVETVYTHVL HPYPTQITQS EKQFVVFEGN HYFYSPYPTK TQTMRVKLAS RNVESYTKLG NPTRSEDLLD YGPFRDVPAY SQDTFKVHYE NNSPFLTITS MTRVIEVSHW GNIAVEENVD LKHTGAVLKG PFSRYDYQRQ PDSGISSIRS F; Positive Samples: HL-60, HepG2, SKOV3, MCF7, BxPC-3, Mouse liver, Rat liver; Cellular Location: Endoplasmic reticulum membrane, Melanosome, Single-pass type I membrane protein
Ribophorins 1 and 2 are abundant and highly conserved glycoproteins residing in the endoplasic reticulum, that participate in ribosome binding. Mammalian oligosaccharyltransferase activity is associated with a protein complex composed of RPN1, RPN2, and an oligosaccharyltransferase protein. RPN1 is a component of the proteasome base. The ubiquitin-like (UBL) domain of recombinant Rad23 interacts with proteasomes through the leucine-rich repeat domain of RPN1. The RPN1 gene maps to chromosome 3 in somatic cell hybrids, and the RPN2 gene maps to chromosome 20 by in situ hybridization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dolichyl-diphosphooligosaccharide--protein; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase 67 kDa subunit; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit 1; dolichyl-diphosphooligosaccharide-protein glycosyltransferase 67 kDa subunit; oligosaccharyltransfe; oligosaccharyltransferase 1 homolog; oligosaccharyltransferase complex subunit (non-catalytic); Ribophorin I; Ribophorin-1; RPN-I
Gene Aliases: AU018702; D6Wsu137e; OST1; RBPH1; RIBI; Rpn-1; RPN-I; RPN1
UniProt ID: (Human) P04843, (Mouse) Q91YQ5
Entrez Gene ID: (Human) 6184, (Rat) 25596, (Mouse) 103963
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support