Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: S Opsin.
Opsin Blue belongs to the G-protein coupled receptor 1 family, opsin subfamily. It encodes the blue cone pigment gene which is one of three types of cone photoreceptors responsible for normal color vision. Defects in this gene are the cause of tritan color blindness (tritanopia). Affected individuals lack blue and yellow sensory mechanisms while retaining those for red and green. Defective blue vision is characteristic.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: blue cone opsin; blue cone opsin-like pigment; Blue cone photoreceptor pigment; blue cone pigment; blue opsin; Blue-sensitive opsin; blue/UV opsin; BOP; opsin 1 (cone pigments), short-wave-sensitive (color blindness, tritan); S opsin; short wavelength sensitive opsin; Short wavelength-sensitive cone opsin; Short-wave-sensitive opsin 1; SWS opsin; UV cone pigment
Gene Aliases: AW551857; BCP; BOP; CBT; Opn1sw
UniProt ID: (Rat) Q63652, (Mouse) P51491
Entrez Gene ID: (Human) 611, (Rat) 81644, (Mouse) 12057
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support