Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
Positive Control:Human esophagus or skin.
Cellular Location: Nucleus. Cytoplasm.
S100A16 (S100 Calcium Binding Protein A16) is a Protein Coding gene. Diseases associated with S100A16 include Robinow Syndrome, Autosomal Recessive and Robinow Syndrome. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity. Calcium-binding protein. Binds one calcium ion per monomer. Can promote differentiation of adipocytes (in vitro). Overexpression in preadipocytes increases their proliferation, enhances adipogenesis and reduces insulin-stimulated glucose uptake.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aging-associated gene 13 protein; aging-associated protein 13; MGC17528; Protein S100-A16; Protein S100-F; S100 calcium-binding protein A16
Gene Aliases: AAG13; DT1P1A7; S100A16; S100F
UniProt ID: (Human) Q96FQ6
Entrez Gene ID: (Human) 140576
Molecular Function: calmodulin-related
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support