Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Control:Monocytes and lymphocytes.
Cellular Location: Nucleus. Cytoplasm.
Binds calcium, zinc and copper. One subunit can simultaneously bind 2 calcium ions or 2 copper ions plus 1 zinc ion. Calcium and copper ions compete for the same binding sites. Engelkamp D., Proc. Natl. Acad. Sci. U.S.A. 90:6547-6551(1993). Gregory S.G., Nature 441:315-321(2006).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein S-100D; Protein S100-A5; S100 calcium-binding protein A5
Gene Aliases: S100A5; S100D
UniProt ID: (Human) P33763
Entrez Gene ID: (Human) 6276
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support